missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561888
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KPQITNPKAE ITRFVPTALR VRRENK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Posttranslational modification of proteins by the addition of the small protein SUMO.Specifica
WBP11 | |
Polyclonal | |
Unconjugated | |
WBP11 | |
2510026P17Rik; D6Wsu113e; DKFZp779M1063; hypothetical protein LOC402950; Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; PPP1R165; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein; SH3 domain-binding protein SNP70; SIPP1; SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; Wbp11; WBP-11; WW domain binding protein 11; WW domain-binding protein 11; zgc:77390 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51729 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2W2 | |
WBP11 | |
Recombinant protein corresponding to Human WBP11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only