Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran
100% pure nitrocellulose to ensure the highest binding capacity possible
Marca: Cytiva 10402495
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
- Also available in circular formats
Specifica
20 cm | |
20cm x 3m roll | |
High | |
Roll | |
0.2 μm |
3 m | |
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection | |
1 Each | |
Nitrocellulose |