Learn More
DWK Life Sciences Wheaton™ Extractors For Soxhlet Extraction Apparatus
For continuous extraction of analytes from a solid into an organic solvent.
Marca: DWK Life Sciences 415010
342.95 EUR valido fino al 2024-12-31
Usa il codice promo "23261" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.90720kg
Descrizione
- Wetted parts are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Specifica
Soxhlet Extractor | |
Borosilicate Glass | |
ASTM E 438 | |
55/50 | |
5 cm | |
25 cm |
Soxhlet Extraction Apparatus | |
Used for continuous extraction of analytes from a solid into an organic solvent | |
24/40 | |
1.96 in. | |
9.84 in. |