Learn More
Spectrum™ Labs White Polypropylene Standard Closures
For standard grade RC dialysis tubing
Marca: Spectrum™ Labs 142734
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
These polypropylene closures lock over the tubing, forming a sack ensuring a leak-proof seal. Regular closures will float in water. They eliminate the need for tying knots in dialysis tubing, a practice which can adversely affect the porosity of a membrane. Lighter than water, regular closures are normally used with weighted or magnetic weighted closures. Attach a regular closure to the top of the dialysis tubing and a weighted or magnetic weighted closure to the bottom for vertical floating. Not for use with cellulose ester membrane tubing. Autoclavable.Specifica
Standard Closure | |
12 mm | |
Polypropylene | |
White | |
0°C to 121°C | |
For standard grade RC dialysis tubing |
Polypropylene | |
Autoclavable | |
Standard Closure | |
Autoclavable | |
10 Pack |