Learn More
Memmert™ Works Calibration Certificate for 3 Temperatures
Factory calibration certificate for 3 temperatures (+5/+37+60°C)
Marca: Memmert™ IPP750ecoD00129
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- +5, +37, +60 °C
- Accessory
- Factory calibration certificate for 3 temperatures
Specifica
IPP750eco | |
Works Calibration Certificate for 3 Temperatures |