Learn More
ABEBA™ X-Light Black Occupational Shoes O2 ESD
Black X-light occupational shoes with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Marca:  ABEBA™ 7131127/42
Dettagli aggiuntivi : Peso : 0.88000kg
Codice prodotto. 17111535
  • 99.71€ / 1 paio
Spedizione stimata: 10-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Lightweight, comfortable black occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Black microfiber
  • Silver ions lining
  • Replaceable active comfort insole
Specifica
Specifica
Black
42
Unisex
Water Resistant
Microfiber
Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Low-cut
O2 SRC ESD