Learn More
Fisherbrand™ Top-Line Pipette Filter Tips
Eliminate sample and pipette contamination. Fisherbrand™ Top-Line Pipette Filter Tips prevent impurities from passing through micropore filter.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.96000kg
Descrizione
- Filters are made of high-quality hydrophobic virgin polyethylene without additives
- Filters block aerosols and liquids through micropore filter technology
- Optimized cone geometry for a secure fit
- Graduation marks on tip at standard volumes (except 0.5-10/20µl filter tip)
- Conical tip outlet prevents drop formation at the outlet opening of tip
- Tips manufactured from autoclavable, non-wettable polypropylene
- Sterile packs available (beta irradiation)
Specifica
1-100 μl | |
Sì | |
Autoclavable | |
Beta radiation | |
Yes | |
Standard Volumes |
Filtro | |
Trasparente | |
5.08 cm | |
Rack | |
Virgin Polypropylene |