Learn More
Chemsplash Xtreme 50 SMS T5/6 Disposable Coverall Red

Descrizione
Chemsplash Xtreme SMS 50 Coveralls
- 50GSM Breathable SMS Fabric
- Elasticated 3 Piece Hood
- Elasticated Cuffs and Ankles
- Two Way Zip with Adhesive Zip Flap
- Silicone and Latex Free
- Comes without the chest label
- PFAS Free
- See Style 2618 for White Sizes 4XL-5XL-6XL
Description: Chemsplash Xtreme SMS 50 disposable coveralls are made from extremely breathable SMS fabric suitable for prolonged periods of use. Featuring an elasticated hood, cuffs, and ankles, with a two-way zip and an adhesive cover flap. Its strong and robust properties provide a good chemical barrier, especially to dry hazardous particles, whilst being extremely comfortable to wear. Offering the best value within our Cat III Type 5 and 6 range. Available in White, Blue, Red, and Orange.
Specifica
Specifica
| Product Type | Hooded Coveralls |
| Size | Small |
| Sesso | Unisex |
| Materiale | SMS Fabric |
| Colore | Red |
| Monouso | Single-use |
| Stile vita | Elastic |
| Ankle Style | Elastic |
| Stile polso | Elastic |
| Stile collo | Hooded |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.