missing translation for 'onlineSavingsMsg'
Learn More

ZBTB7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11341-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898343

  • 349.00€ / 150µL

Spedizione stimata: 23-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a zinc finger-containing transcription factor that acts as a key regulator of lineage commitment of immature T-cell precursors. It is necessary and sufficient for commitment of CD4 lineage, while its absence causes CD8 commitment. It also functions as a transcriptional repressor of type I collagen genes. Alternatively spliced transcript variants have been found for this gene.
Specifica

Specifica

ZBTB7B
Polyclonal
Unconjugated
ZBTB7B
c Krox, hcKrox, THPOK, ZBTB15, ZBTB7B, Zfp 67, ZFP67, Zinc finger protein 67 homolog, Zinc finger protein Th POK, ZNF857B
Rabbit
Antigen Affinity Chromatography
RUO
22724, 295248, 51043
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15156, Q64321
ZBTB7B
ZBTB7B Fusion Protein Ag1888
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali