Learn More
3M™ Zeta Plus™ EXT Series Filter Cartridge with SP Series Media
3M™ Zeta Plus™ EXT Series Filter Cartridge with SP Series Media is a dual layer depth filter composed of inorganic filter aid, cellulose and a positively charged resin system that pulls negatively charged contaminants from the fluid.
Marca: 3M™ PU0205ZP
Dettagli aggiuntivi : Peso : 8.90000kg
Descrizione
- Double layers of complementary porosities provide high contaminant holding capacity, economical filtration and reliable particle reduction.
- Enhanced protection of downstream membrane filters.
- A complete particle retention range from coarse to fine.
- Open grade EXT filters are designed for primary clarification or to replace centrifuges and TFF systems.
- Consolidation of multiple filter stages resulting in lower processing costs.
- FDA Drug Master File and USP Class VI Biological Reactivity Tests eases validation and regulatory submissions.
Specifica
Z16E08AA60SP02A |