Learn More
3M™ Zeta Plus™ EXT Series Filter Cartridge with SP Series Media
3M™ Zeta Plus™ EXT Series Filter Cartridge with SP Series Media is a dual layer depth filter composed of inorganic filter aid, cellulose and a positively charged resin system that pulls negatively charged contaminants from the fluid.
Marca:  3M™ PU0205ZP
Dettagli aggiuntivi : Peso : 8.90000kg
Codice prodotto. 16661156
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
  • Double layers of complementary porosities provide high contaminant holding capacity, economical filtration and reliable particle reduction.
  • Enhanced protection of downstream membrane filters.
  • A complete particle retention range from coarse to fine.
  • Open grade EXT filters are designed for primary clarification or to replace centrifuges and TFF systems.
  • Consolidation of multiple filter stages resulting in lower processing costs.
  • FDA Drug Master File and USP Class VI Biological Reactivity Tests eases validation and regulatory submissions.
Specifica
Specifica
Z16E08AA60SP02A