Learn More
ZNF583 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554890
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EPWMVKKEGT RGPCPDWEYV FKNSEFSSKQ ETYEESSKVV TVGARHLSYS LDYPSLREDC QSEDWYKNQL GSQEVH Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 55%.
ZNF583 may be involved in transcriptional regulation.Specifica
ZNF583 | |
Polyclonal | |
Unconjugated | |
ZNF583 | |
BC056221; LOC101788406; LOW QUALITY PROTEIN: zinc finger protein 583; RGD1562044; Zfp583; Zinc finger protein 583; zinc finger protein L3-5; Znf583 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96ND8 | |
ZNF583 | |
Recombinant protein corresponding to Human ZNF583 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only