missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19426-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.Specifica
ZNF593 | |
Polyclonal | |
Unconjugated | |
ZNF593 | |
zinc finger protein 593, Zinc finger protein T86, ZNF593, ZT86 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51042, 68040 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00488, Q9DB42 | |
ZNF593 | |
ZNF593 Fusion Protein Ag13758 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |