CAS RN 10080-05-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
4
di
4
risultati
1
1-Cyclohexyl-3-(p-sulfamoylphenethyl)urea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Cyclohexyl-3-(p-sulfamoylphenethyl)urea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
1-Cyclohexyl-3-(p-sulfamoylphenethyl)urea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.