CAS RN 35748-51-7
CAS RN 35748-51-7
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
MedChemExpress Pressinoic Acid
MedChemExpress Pressinoic Acid is a synthetic hexapeptide with potent corticotrophin-releasing activity. Pressinoic Acid is also an oxytocin inhibitor; it induces maternal behavior.
2
MedChemExpress Pressinoic Acid
MedChemExpress Pressinoic Acid is a synthetic hexapeptide with potent corticotrophin-releasing activity. Pressinoic Acid is also an oxytocin inhibitor; it induces maternal behavior.
3
MedChemExpress Pressinoic Acid
MedChemExpress Pressinoic Acid is a synthetic hexapeptide with potent corticotrophin-releasing activity. Pressinoic Acid is also an oxytocin inhibitor; it induces maternal behavior.