CAS RN 58-08-2
Caffeine, 98.5%, specified according to the req. of USP/BP
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 IUPAC Name: 1,3,7-trimethylpurine-2,6-dione SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine, 99.7%
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 IUPAC Name: 1,3,7-trimethylpurine-2,6-dione SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine, 99%
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine Anhydrous MP Biomedicals
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 IUPAC Name: 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine monohydrate, MP Biomedicals™
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 IUPAC Name: 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine anhydrous, 99.7%, MP Biomedicals™
CAS: 58-08-2 Formula molecolare: C8H10N4O2 Molecular Weight (g/mol): 194.19 Numero MDL: MFCD00005758 InChI Key: RYYVLZVUVIJVGH-UHFFFAOYSA-N Sinonimo: caffeine,1,3,7-trimethylxanthine,guaranine,thein,cafeina,methyltheobromine,koffein,mateina,theine,alert-pep PubChem CID: 2519 ChEBI: CHEBI:27732 IUPAC Name: 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione SMILES: CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Caffeine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Caffeine 1.0 mg/ml in Methanol, LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Caffeine, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Caffeine, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Pharmaceuticals Mixture 326 200 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Caffeine, TRC
CAS: 58-08-2 Formula molecolare: C8 H10 N4 O2 Molecular Weight (g/mol): 194.19 Sinonimo: Caffeine (8CI),3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione,1,3,7-Trimethyl-2,6-dioxopurine,1,3,7-Trimethyl-7H-purine-2,6-dione,1,3,7-Trimethylxanthine,7-Methyltheophylline,Alert-Pep,Asia migrine,Cafalgine,Cafeina,Caffedrine,Caffein,Cafipel,DHCplus,Dasin,Diurex,Durvitan,Guaranine,Koffein,Mateina,Methyltheobromine,Midron extra,Miudol,NSC 5036,New Cetamol,No-Doz,Palergot-C,Phensal,Refresh'n,Shape Plus,Stay Alert,Stim,Synalgos,Thein,Theine,Tri-Aqua,Vivarin,Wigraine IUPAC Name: 1,3,7-trimethylpurine-2,6-dione SMILES: CN1C(=O)N(C)c2ncn(C)c2C1=O
- 1
- 2