Cell Culture Microplates
Cell culture microplates are essential consumables for in vitro cell growth, analysis, and screening across research, pharmaceutical, and biotechnology laboratories. Designed to support consistent cell attachment and proliferation, these plates are available in multiple well formats, surface treatments, and optical properties to meet diverse experimental needs. Discover reliable solutions from leading brands to support reproducible and scalable cell culture workflows.
Useful Links
Save Now - Exclusive Deals
Product Code 10119831
Product Code 10604903
Product Code 10695951
Product Code 10078850
Product Code 10360691
Product Code 10344311
Product Code 10098870
Product Code 10334791
Product Code 10167481
Complete Your Order - Great Deals
Must Have
Product Code 10334791
Product Code 10098870
Product Code 10344311
Product Code 10167481
FAQ
Cell culture microplates are used to grow, maintain, and analyze cells in controlled in vitro environments. They support applications such as drug screening, toxicity testing, and cell-based assays using brands like Corning and Thermo Scientific Nunc.
The correct well format depends on throughput and assay volume. 96- and 384-well plates support high-throughput screening, while 6- and 24-well plates are ideal for routine cell culture and microscopy.
TC-treated and Nunclon Delta surfaces enhance cell attachment and growth. These treatments are commonly used for adherent cell lines and are offered by Corning, Thermo Scientific Nunc, and Fisherbrand.
Optical clarity enables accurate imaging and fluorescence or luminescence detection. Optical-bottom plates from Corning and Thermo Scientific are designed to improve signal quality in advanced assays.
All listed microplates have Accessibility = 15, meaning they are set up for use and availability across all supported European countries.