UserName
General Clinical Specimen Containers
General Clinical Specimen Containers
Specialized storage containers designed to collect, hold, transport, and stabilize general clinical specimens. Products are available in several sizes, capacities, and material compositions.
Thermo Scientific Sterilin Polystyrene Containers, 60mL to 250mL
Store samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities.
Thermo Scientific™ Sterilin™ Polystyrene Containers, 60mL to 250mL
Sustainable General Clinical Specimen Containers
Safely collect and transport samples for lab analysis.
All Special Offers on General Clinical Specimen Containers
Stainless Steel General Clinical Specimen Containers
Durable and resistant to contamination, stainless steel containers are ideal for secure sample collection and transport.
Stainless Steel General Clinical Specimen Containers
Complete Your Order - Great Deals
Fisherbrand High Precision, 45° Curved, Tapered, Very Fine Point Tweezers/Forceps
Allow maximum visibility under magnified applications. Fisherbrand™ High Precision, 45° Curved, Tapered, Very Fine Point Tweezers/Forceps are made of anti-magnetic stainless steel.
Electronic Toploading Balances
With a higher capacity than analytical balances, toploading balances are designed to weigh quantities up to the kg level. With a wide choice in weighing capacity and measurement precision, internal or external calibration systems and integrated software are available according to your needs.
Exam Gloves
Examination gloves are made from a variety of materials including latex, nitrile, polyvinyl chloride, and other polymers. In addition to their use in clinical or healthcare settings, examination gloves are also worn to protect the hands in chemical and biochemical laboratories, as well as in manufacturing and controlled environments.
FAQ
A clinical specimen container is a specially designed vessel used to collect, store, and transport biological samples from patients for diagnostic testing, research, or clinical analysis. These containers are crucial for maintaining the integrity of the specimen and preventing contamination. They come in various types depending on the specimen to be collected:
  • Urine containers: Sterile cups or tubes for collecting urine samples
  • Blood collection tubes: Vacuum-sealed tubes with specific additives for blood collection
  • Swabs: For collecting samples from the throat, nasal passages, or wounds
  • Stool containers: Designed for the collection and transport of fecal samples
  • Sputum containers: For collecting mucus from the respiratory tract
Clinical specimen containers are usually labeled with patient information, date, and time of collection to ensure accurate identification and traceability.
To handle a clinical specimen container properly:
  • Wear PPE: Gloves, lab coat, and eye protection
  • Use aseptic techniques: Ensure the container is sterile
  • Collect sample: Follow specific procedures for the type of specimen
  • Seal container: Securely close to prevent leaks
  • Label accurately: Include patient info, date, and time
  • Transport safely: Use leak-proof bags and follow guidelines
  • Store appropriately: If not transported immediately, follow storage guidelines
  • Dispose properly: Use biohazard containers for waste
These steps ensure sample integrity and prevent contamination.
When selecting general clinical specimen containers, consider:
  • Sterility: Ensure the container is sterile to prevent contamination
  • Material: Choose durable, non-reactive materials like plastic
  • Size: Select appropriate size for the volume of specimen needed
  • Sealability: Ensure a secure lid to prevent leaks
  • Labeling: Look for containers with adequate labeling space for patient information
Shop By Brand