Learn More
Bochem™ 18/10 Stainless Ladle Scoops with Flat Handle
Marca: Bochem™ 8794
Dettagli aggiuntivi : Peso : 0.50000kg
Spécification
Ladle Scoop | |
Flat | |
400 mm |
Stainless Steel (18/10) | |
250 mL |