Learn More
Linker Industrietechnik™ 50 Dia. x 25mm Height Stainless Steel Test Sieves
Marca: Linker Industrietechnik™ 550.025.222062
Dettagli aggiuntivi : Peso : 0.80000kg
Specifica
Stainless Steel | |
50 mm | |
2mm | |
Compliance 2.1 EN 10 204 and ISO 3310-1 | |
25mm |
8211 | |
1Each | |
Test Sieve | |
Stainless Steel |