Learn More
Spectrum™ Labs 8 - 10 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Marca: Spectrum™ 131270T
Dettagli aggiuntivi : Peso : 0.01000kg
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Descrizione
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifica
Dialysis Trial Kit | |
Cellulose Ester | |
0.39 in. | |
1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks | |
Ideal for small, critical dialysis applications |
8 to 10 kDa | |
10 mm | |
0.79 mL/cm vol./length | |
Each |