Learn More
Uvex™ 9770 Pheos E Unventilated Safety Helmets
Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. A conventional 6-point textile strap interior allows for easy adjustment.
Marca:  Uvex™ 9770220
Dettagli aggiuntivi : Peso : 0.40100kg
Codice prodotto. 17219101
  • 12.65€ / Pezzo
Spedizione stimata: 05-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Lightweight, unventilated helmet in a sporty design
  • Completely closed for use in the electrician sector with low voltage systems
  • Includes an adapter system for visors, helmet caps, and helmet lamps
  • Conventional 6-point textile strap interior allows for easy adjustment
  • Style 9770026 is pasted with the highly reflective pheos reflex sticker set
Specifica
Specifica
EN 397:2012, EN 50365:2002
Low Voltage Systems
HDPE
Electrically Insulating Helmet
Not Ventilated
Orange
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps
9770 Pheos E
52 to 61 cm