Learn More
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes with these accessories, including storage bottles and electrode bulb guards.
Marca: Thermo Scientific™ 020017
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
- Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
- Designed for glass body pH electrodes
- Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
Specifica
Electrode Storage Chamber | |
No Cal Electrode | |
Electrode Storage Chamber |