Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
Marca: Cytiva (formerly Pall Lab) AP-4794
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
- For research use only
Specifica
Glass Fiber, PVDF | |
Non-sterile | |
<125 μL | |
1000/Pk. | |
25 mm |
0.2 μm | |
3.9 cm2 | |
Polypropylene | |
82°C |