Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 96 Filter Plates, 1 mL
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Cytiva (formerly Pall Lab) 5055
Dettagli aggiuntivi : Peso : 0.58900kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
PTFE | |
0.2 μm | |
5/Pk. |
PTFE | |
Non-sterile | |
900 μL (Sample) |