Learn More
Pall AcroPrep™ Advance 96-well Filter Plates for Protein Purification
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Pall 8039
241.90 EUR valido fino al 2023-02-05
Usa il codice promo "19086" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.12100kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Supor (PES) | |
Non-sterile | |
10/Pk. |
1.2 μm | |
Polypropylene Housing | |
350 μL |