Learn More
FTM Technologies™ Active Carbon Filters
Used to eliminate noxious or unpleasant smells emanating from liquids, vapours or smoke. FTM Technologies™ Active Carbon Filters are for use with fume hoods.
Marca: FTM Technologies™ FC2
Dettagli aggiuntivi : Peso : 0.65700kg
Descrizione
- Superior protection in the laboratory
- Compatible with a wide range of fume hoods
Specifica
Per 27919/57 | |
Per vapori organici, acidi e odori | |
Filtro a carbone attivo |
Activated Carbon | |
1 ciascuno |