Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
Marca: Eppendorf™ 5427733000
Dettagli aggiuntivi : Peso : 0.12000kg
Descrizione
- Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifica
Large Rotor Bore Adapter | |
1 x 2.6 to 7 mL Tube | |
Large rotor bore |
Round Bottom, Blood Collection Tubes | |
Rotor F-35-6-30 | |
2/Pk. |