missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin Mouse, Clone: ADPN/1370, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 9370-MSM3-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Adiponectin | |
Cytoplasmic and Cell Surface | |
ADPN/1370 | |
Unconjugated | |
NP_001171271 | |
Mouse | |
26-30kDa | |
Protein A/G Purified | |
RUO | |
9370 | |
IgG2b |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ADIPOQ | |
Recombinant human Adiponectin protein fragment | |
HeLa cells or Kidney | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |