missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPRH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17911-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Catalyzes the reverse reaction of mono-ADP-ribosylation.Specifica
ADPRH | |
Polyclonal | |
Unconjugated | |
ADPRH | |
ADP ribosylarginine hydrolase, ADPRH, ARH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11544, 141, 25371 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54922, P54923, Q02589 | |
ADPRH | |
ADPRH Fusion Protein Ag12311 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |