missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHNAK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17682-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The specific function of AHNAK2 is not yet known.Specifica
AHNAK2 | |
Polyclonal | |
Unconjugated | |
AHNAK2 | |
AHNAK nucleoprotein 2, AHNAK2, C14orf78, KIAA2019, Protein AHNAK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113146 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IVF2 | |
AHNAK2 | |
AHNAK2 Fusion Protein Ag11779 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |