missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A2 Mouse anti-Human, Clone: 2E1E8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66677-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.Specifica
AKR7A2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43488 | |
AKR7A2 | |
AKR7A2 Fusion Protein Ag26105 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2E1E8 | |
Unconjugated | |
AKR7A2 | |
AFAR, AFAR1, AFB1 aldehyde reductase 1, AFB1 AR 1, AFB1 AR1, AKR7, AKR7A2, Aldoketoreductase 7, SSA reductase | |
Mouse | |
Protein G | |
RUO | |
8574 | |
-20°C | |
Liquid |