Learn More
Al-23 Tube, Both Ends Open, O.D.: 24mm, I.D.: 18mm, Thermo Scientific Chemicals
Marca:  Thermo Scientific Alfa Aesar 031572.DW
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 11333997
  • 187.00€ / 300mm
Spedizione stimata: 14-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
Al-23 Tube
Both ends open
300 mm