Learn More
Emma Safety Footwear Alaska XXD Safety Shoes
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
Marca: Emma Safety Footwear MM30603639
Dettagli aggiuntivi : Peso : 1.75000kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifica
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
39 |
Laces | |
Alaska XXD | |
Oiled Nubuck | |
Steel |