Learn More
Ansell AlphaTec™ 5000
This highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
Marca:  Ansell OR50T-00103-04
Dettagli aggiuntivi : Peso : 6.50000kg
Codice prodotto. 17110865
  • 767.37€ / pezzi 10
Spedizione stimata: 25-05-2023
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Designed to protect, rear entry suit with neoprene rubber face seal for a close fit to full face respirators
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Designed to protect, typical innovative features include dual zip systems, double cuffs with knitted inner cuffs
  • Multi-layer barrier material which is lightweight, yet strong and durable
  • Anti-static
Specifica
Specifica
Large
Orange
Welded and Taped
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases
Multi-Layer Non-Woven Barrier Laminate Fabric
O50T103
EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
132 gsm