Learn More
Abeba™ Anatom 2120 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca:  Abeba™ 2120/40
Dettagli aggiuntivi : Peso : 2.50000kg
Codice prodotto. 15362868
  • 101.70€ / 1 paio
Spedizione stimata: 05-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • Suitable for heel and foot lift
Specifica
Specifica
CE EN ISO 20345:2011 SB A, E SRC
Microfiber
Slip-resistant
2120
White
40
Polyurethane
Steel