missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18302-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.Specifica
Angiogenin | |
Polyclonal | |
Unconjugated | |
ANG | |
ALS9, ANG, Angiogenin, HEL168, Ribonuclease 5, RNase 5, RNASE4, RNASE5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
283 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P03950 | |
ANG | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |