missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human CD56, Clone: NCAM16.2; BV421 Conjugate BD
Marca: BD Biosciences 562752
Dettagli aggiuntivi : Peso : 0.00800kg
Descrizione
Host Species: Mouse
Clone: NCAM16.2
Isotype: IgG2b, κ
Cross Reactivity Species: Hu
Immunogen: Immunoaffinity-enriched adult human brain NCAM
Flow Cytometry
Specifica
Human CD56 | |
NCAM16.2 | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Monoclonal | |
Brilliant Violet 421 | |
Immunoaffinity-enriched adult human brain NCAM | |
25 Tests | |
Primary |