Learn More
anti-IgG1 κ, Rat, FITC, Clone: R3-34, Isotype Control, BD
Marca: BD Biosciences 553924
358.59 EUR valido fino al 2024-05-31
Usa il codice promo "21638" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Flow Cytometry, Intracellular Staining, Isotype Control
Specifica
Flow Cytometry, Western Blot | |
0.5mg/mL | |
Rat | |
0.25 mg | |
IgG1 κ |
R3-34 | |
FITC | |
Affinity Purified | |
Undiluted at 4°C |