missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein H Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4D9A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66074-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.Specifica
Apolipoprotein H | |
Monoclonal | |
0.78 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02749, P26644, Q01339 | |
Apoh | |
Apolipoprotein H Fusion Protein Ag17915 | |
20 μL | |
Primary | |
Rat, Porcine, Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
4D9A4 | |
Unconjugated | |
APOH | |
Anticardiolipin cofactor, APC inhibitor, Apo H, APOH, Apolipoprotein H, B2G1, B2GPI, Beta 2 glycoprotein 1, Beta 2 glycoprotein I, Beta(2)GPI, BG | |
Mouse | |
Protein G | |
RUO | |
100522100, 11818, 287774, 350 | |
-20°C | |
Liquid |