missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMCX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20193-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintece of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome.Specifica
ARMCX1 | |
Polyclonal | |
Unconjugated | |
ARMCX1 | |
ALEX1, ARMCX1, Protein ALEX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51309, 78248 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CX83, Q9P291 | |
ARMCX1 | |
ARMCX1 Fusion Protein Ag14120 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |