Learn More
Thermo Scientific™ Avermectin, ACROS Organics™
Marca: Thermo Scientific™ 468392500
Dettagli aggiuntivi : CAS : 65195-55-3 Peso : 0.00025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : I
Descrizione
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Avermectin | |
250 mg | |
873.09 | |
White to Light-yellow |
65195-55-3 | |
C48 H72 O14 | |
Crystalline Powder |