missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AXUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18162-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Axin, an important regulator of beta-catenin, is frequently mutated in human hepatocellular carcinomas (HCCs), and transduction of the wild-type Axin gene (AXIN1) induces apoptosis in HCC cells as well as in colon cancer cells. A novel gene, AXUD1 (AXIN1 up-regulated), has enhanced expression in response to exogenously expressed AXIN1. It is expressed in all human tissues examined, most abundantly in lung, placenta, skeletal muscle, pancreas and leukocytes. Data suggests that AXUD1 may have a tumor-suppressor function in those organs.Specifica
AXUD1 | |
Polyclonal | |
Unconjugated | |
CSRNP1 | |
AXIN1 up regulated 1, AXUD1, CSRNP1, CSRNP1 Ens, FAM130B, TAIP 3, URAX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215418, 363165, 64651 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59054, Q96S65 | |
Csrnp1 | |
AXUD1 Fusion Protein Ag6158 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |