Learn More
Axygen™ Axypet-16 Pipette Tips
For 1536-well pipetting
Marca: Axygen™ T-1536-L-R-S
Dettagli aggiuntivi : Peso : 10.40000kg
Descrizione
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Maxymum Recovery technology for ultra-low retention and high yield
- Sterile
Specifica
0.5 to 30 μL | |
Clear | |
For 1536-well pipetting | |
Low Retention | |
1536-Well Pipetting |
Sterile | |
DNase, RNase Free | |
Rack | |
384 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polypropylene |