Learn More
Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems
Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems are constructed of polypropylene, and feature a variety of available options to suit your needs.
Marca: Axygen™ RRP-200-C-R
Dettagli aggiuntivi : Peso : 11.30000kg
Description
- Available with or without aerosol-resistant filters
- Available sterile/non-sterile
Specifications
200 μL | |
Non-sterile | |
RNase-free, DNase-free, human gDNA-free, Nonpyrogenic | |
Automation, Round | |
Automation Micropipette Tip | |
Tip |
Non-Filtered | |
Clear | |
Rack | |
10 racks/pk, 5 pk/cs | |
PerkinElmer MultiProbe | |
Polypropylene |