Learn More
Brady™ Nastro in vinile in rotolo continuo per stampanti BBP3x
Labels constructed of B-7569 Vinyl for indoor/outdoor use.
Marca: Brady™ 120923
Dettagli aggiuntivi : Peso : 2.50000kg
Descrizione
- B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels, and visual workplace labels
Specifica
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
30 m x 75 mm |
White | |
Vinyl (B-7569) | |
BBP30, BBP31, BBP33, BBP35, BBP37, i3300, i5300, S3000, S3100 |