missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18098-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein and contains a signal anchor that is not cleaved. It prefers the substrates of lacto-N-tetraose and lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains and the biosynthesis of the backbone structure of dimeric sialyl Lewis a. It plays domit roles in L-selectin ligand biosynthesis, lymphocyte homing and lymphocyte trafficking.Specifica
B3GNT3 | |
Polyclonal | |
Unconjugated | |
B3GNT3 | |
B3GAL T8, B3GALT8, B3GN T3, B3GNT 3, B3GNT3, Beta 1,3 GalTase 8, Beta 1,3 Gn T3, Beta 3 Gx T8, Beta3Gal T8, Beta3GalT8, beta3Gn T3, BGnT 3, Core1 beta3GlcNAcT, HP10328, TMEM3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10331, 72297 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5JCS9, Q9Y2A9 | |
B3gnt3 | |
B3GNT3 Fusion Protein Ag12061 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |