Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps.
Marca: Thermo Scientific™ 09.4002
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
Lampade UV di ricambio per i sistemi di purificazione idrica nel punto di utilizzo Thermo Scientific™ Barnstead™.
Ideal for use with the following Barnstead point of use water purification systems:09.1002:
- MicroPure™ (UV and UV/UVF systems only)
- Smart2Pure™ (UV and UV/UVF systems only)
09.2002:
- GenPure™ (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- GenPure™ Pro (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- GenPure™ xCAD (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- LabTower™ EDI
09.4002:
- Pacific™ TII
- LabTower™ TII
09.5002:
For storage reservoirs (230V only) used with the following Barnstead water purification systems:
- LabTower RO
- LabTower TII
- Pacific TII (30 L, 60 L, 100 L only)
- Smart2Pure (12 only)
Specifica
Replacement UV lamp, for use with Pacific TII, LabTower TII | |
Pacific TII; LabTower TII | |
UV Lamp |