missing translation for 'onlineSavingsMsg'
Learn More

BCL10 Mouse anti-Human, Clone: 2E8G5, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66556-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898535

  • 325.38€ / 150µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifica

Specifica

BCL10
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95999
BCL10
BCL10 Fusion Protein Ag12162
150 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
2E8G5
Unconjugated
BCL10
B cell CLL/lymphoma 10, B cell lymphoma/leukemia 10, BCL10, c E10, CARD like apoptotic protein, CARMEN, cCARMEN, Cellular E10, Cellular homolog of vCARMEN, CIPER, CLAP, hCLAP, mE10
Mouse
Protein G
RUO
8915
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali