missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Lauryl Sulfate Broth
For detection of coliform bacteria
Marca: BD 211338
Dettagli aggiuntivi : Peso : 0.53060kg
Specifica
AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW | |
Dehydrated | |
Lauryl Sulfate Broth | |
Pancreatic digest of Casein 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L | |
Bottle |
Refrigerate | |
Lauryl Sulfate Broth | |
Lauryl Sulfate broth | |
Detection of Coliform bacteria | |
500 g |