missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Rat Anti-Human CXCR5 (CD185)

In peripheral blood, CXCR5 expression is restricted to B lymphocytes and a small subset of CD4+ and CD8+ lymphocytes. BD Horizon™ BV480 Rat Anti-Human CXCR5 (CD185) has restricted expression pattern of CXCR5 on B cells and follicular T helper cells (Tfh) that suggests this receptor functions as a regulator of B and T cell migration.

Marca:  BD 566191

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15844829

  • 316.00€ / Pezzo
Spedizione stimata: 07-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifica

Specifica

BV480 Rat Anti-Human CXCR5 (CD185)
Flow Cytometry
BV480
Store undiluted at 4°C
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
5 μL
QC Testing: Human
5 μL
25 Tests
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali